Learn More
Biolegend PE anti-human CD15 (SSEA-1)
Supplier:  Biolegend 301905
Encompass_Preferred
PE anti-human CD15 (SSEA-1) [HI98]; Isotype: Mouse IgM, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-165-774