Learn More
Biolegend PE anti-human CD150 (SLAM)
Supplier:  Biolegend 306308
Encompass_Preferred
PE anti-human CD150 (SLAM) [A12 (7D4)]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-337