Learn More
Biolegend PE anti-human CD155 (PVR)
Supplier:  Biolegend 337508
Encompass_Preferred
PE anti-human CD155 (PVR) [TX24]; Isotype: Mouse IgG2a, λ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-917