Learn More
Biolegend PE anti-human CD166
Supplier:  Biolegend 343903
Encompass_Preferred
PE anti-human CD166 [3A6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-168-222