Learn More
Biolegend PE anti-human CD172b (SIRPβ)
Supplier:  Biolegend 323906
Encompass_Preferred
PE anti-human CD172b (SIRPβ) [B4B6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC; Size: 100 tests
Catalog No. 50-167-209