Learn More
Biolegend PE anti-human CD179b (Ig λ5)
Supplier:  Biolegend 349804
Encompass_Preferred
PE anti-human CD179b (Ig λ5) [HSL11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC; Size: 100 tests
Catalog No. 50-168-550