Learn More
Biolegend PE anti-human CD182 (CXCR2)
Supplier:  Biolegend 320705
Encompass_Preferred
PE anti-human CD182 (CXCR2) [5E8/CXCR2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-031