Learn More
Biolegend PE anti-human CD183 (CXCR3)
Supplier:  Biolegend 353705
Encompass_Preferred
PE anti-human CD183 (CXCR3) [G025H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-168-805