Learn More
Biolegend PE anti-human CD195 (CCR5) Phospho (Ser349)
Supplier:  Biolegend 321606
Encompass_Preferred
PE anti-human CD195 (CCR5) Phospho (Ser349) [E11/19]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-167-101