Learn More
Biolegend PE anti-human CD205 (DEC-205)
Supplier:  Biolegend 342203
Encompass_Preferred
PE anti-human CD205 (DEC-205) [HD30]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-168-113