Learn More
Biolegend PE anti-human CD207 (Langerin)
Supplier:  Biolegend 352204
Encompass_Preferred
PE anti-human CD207 (Langerin) [10E2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-168-703