Learn More
Biolegend PE anti-human CD21
Supplier:  Biolegend 354904
Encompass_Preferred
PE anti-human CD21 [Bu32]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-403-516