Learn More
Biolegend PE anti-human CD210 (IL-10 R)
Supplier:  Biolegend 308804
Encompass_Preferred
PE anti-human CD210 (IL-10 R) [3F9]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-487