Learn More
Biolegend PE anti-human CD215 (IL-15Rα)
Supplier:  Biolegend 330207
Encompass_Preferred
PE anti-human CD215 (IL-15Rα) [JM7A4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-555