Learn More
Biolegend PE anti-human CD218a (IL-18Rα)
Supplier:  Biolegend 313808
Encompass_Preferred
PE anti-human CD218a (IL-18Rα) [H44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-791