Learn More
Biolegend PE anti-human CD22
Supplier:  Biolegend 302506
Encompass_Preferred
PE anti-human CD22 [HIB22]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. 50-165-871