Learn More
Biolegend PE anti-human CD220
Supplier:  Biolegend 352603
Encompass_Preferred
PE anti-human CD220 [B6.220]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-168-725