Learn More
Biolegend PE anti-human CD221 (IGF-1R)
Supplier:  Biolegend 351806
Encompass_Preferred
PE anti-human CD221 (IGF-1R) [1H7/CD221]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-685