Learn More
Biolegend PE anti-human CD25
Supplier:  Biolegend 302605
Encompass_Preferred
PE anti-human CD25 [BC96]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-165-878