Learn More
Biolegend PE anti-human CD255 (TWEAK)
Catalog No. 50166474
Encompass_Preferred
PE anti-human CD255 (TWEAK) [CARL-1]; Isotype: Mouse IgG3, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 25 tests
Catalog No. 50-166-474 Supplier Biolegend Supplier No. 308305
PE anti-human CD255 (TWEAK) [CARL-1]; Isotype: Mouse IgG3, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 25 tests