Learn More
Biolegend PE anti-human CD26
Supplier:  Biolegend 302706
Encompass_Preferred
PE anti-human CD26 [BA5b]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-165-893