Learn More
Biolegend PE anti-human CD266 (Fn14, TWEAK Receptor)
Supplier:  Biolegend 314004
Encompass_Preferred
PE anti-human CD266 (Fn14, TWEAK Receptor) [ITEM-1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-166-797