Learn More
Biolegend PE anti-human CD275 (B7-H2, B7-RP1, ICOSL)
Supplier:  Biolegend 329805
Encompass_Preferred
PE anti-human CD275 (B7-H2, B7-RP1, ICOSL) [9F.8A4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-516