Learn More
Biolegend PE anti-human CD276 (B7-H3)
Supplier:  Biolegend 331606
Encompass_Preferred
PE anti-human CD276 (B7-H3) [DCN.70]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-676