Learn More
Biolegend PE anti-human CD282 (TLR2)
Supplier:  Biolegend 309707
Encompass_Preferred
PE anti-human CD282 (TLR2) [TL2.1]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-166-515