Learn More
Biolegend PE anti-human CD300e (IREM-2, CMRF35-A5)
Supplier:  Biolegend 339704
Encompass_Preferred
PE anti-human CD300e (IREM-2, CMRF35-A5) [UP-H2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-011