Learn More
Biolegend PE anti-human CD307d (FcRL4)
Supplier:  Biolegend 340204
Encompass_Preferred
PE anti-human CD307d (FcRL4) [413D12]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-037