Learn More
Biolegend PE anti-human CD314 (NKG2D)
Supplier:  Biolegend 320805
Encompass_Preferred
PE anti-human CD314 (NKG2D) [1D11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-039