Learn More
Biolegend PE anti-human CD32
Supplier:  Biolegend 303206
Encompass_Preferred
PE anti-human CD32 [FUN-2]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-165-974