Learn More
Biolegend PE anti-human CD334 (FGFR4)
Supplier:  Biolegend 324305
Encompass_Preferred
PE anti-human CD334 (FGFR4) [4FR6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-242