Learn More
Biolegend PE anti-human CD344 (Frizzled-4)
Supplier:  Biolegend 326605
Encompass_Preferred
PE anti-human CD344 (Frizzled-4) [CH3A4A7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-167-334