Learn More
Biolegend PE anti-human CD35
Supplier:  Biolegend 333405
Encompass_Preferred
PE anti-human CD35 [E11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-716