Learn More
Biolegend PE anti-human CD4
Supplier:  Biolegend 344605
Encompass_Preferred
PE anti-human CD4 [SK3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-168-260