Learn More
Biolegend PE anti-human CD54
Supplier:  Biolegend 353106
Encompass_Preferred
PE anti-human CD54 [HA58]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-748