Learn More
Biolegend PE anti-human CD56 (NCAM)
Supplier:  Biolegend 362507
Encompass_Preferred
PE anti-human CD56 (NCAM) [5.1H11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-850-815