Learn More
Biolegend PE anti-human CD79a (Igα)
Supplier:  Biolegend 333504
Encompass_Preferred
PE anti-human CD79a (Igα) [HM47]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-167-720