Learn More
Biolegend PE anti-human CD8
Supplier:  Biolegend 344706
Encompass_Preferred
PE anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: African Green, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 100 tests
Catalog No. 50-168-280