Learn More
Biolegend PE anti-human CD86
Supplier:  Biolegend 305405
Encompass_Preferred
PE anti-human CD86 [IT2.2]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Common Marmoset, Cotton-topped Tamarin, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-255