Learn More
Biolegend PE anti-human CD370 (CLEC9A/DNGR1)
Supplier:  Biolegend 353804
Encompass_Preferred
PE anti-human CD370 (CLEC9A/DNGR1) [8F9]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-818