Learn More
Biolegend PE anti-human CX3CR1
Supplier:  Biolegend 355704
Encompass_Preferred
PE anti-human CX3CR1 [K0124E1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-604-416