Learn More
Biolegend PE anti-human Delta Opioid Receptor
Supplier:  Biolegend 327206
Encompass_Preferred
PE anti-human Delta Opioid Receptor [DOR7D2A4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-5750