Learn More
Biolegend PE anti-human FcεRIα
Supplier:  Biolegend 334609
Encompass_Preferred
PE anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-797