Learn More
Biolegend PE anti-human HLA-DRB1
Supplier:  Biolegend 362304
Encompass_Preferred
PE anti-human HLA-DRB1 [NFLD.D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-217