Learn More
Biolegend PE anti-human HLA-E
Supplier:  Biolegend 342604
Encompass_Preferred
PE anti-human HLA-E [3D12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-139