Learn More
Biolegend PE anti-human IL-10
Supplier:  Biolegend 501403
Encompass_Preferred
PE anti-human IL-10 [JES3-9D7]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Rhesus, Cynomolgus; Apps: ICFC; Size: 25 tests
Catalog No. 50-169-673