Learn More
Biolegend PE anti-human IL-12/IL-23 p40
Supplier:  Biolegend 501807
Encompass_Preferred
PE anti-human IL-12/IL-23 p40 [C11.5]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICFC; Size: 100 tests
Catalog No. 50-169-696