Learn More
Biolegend PE anti-human IL-13
Supplier:  Biolegend 501903
Encompass_Preferred
PE anti-human IL-13 [JES10-5A2]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-169-704