Learn More
Biolegend PE anti-human IL-1α
Supplier:  Biolegend 500106
Encompass_Preferred
PE anti-human IL-1α [364-3B3-14]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: ICFC; Size: 100 tests
Catalog No. 50-169-588