Learn More
Biolegend PE anti-human IL-9
Supplier:  Biolegend 507604
Encompass_Preferred
PE anti-human IL-9 [MH9A4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-170-031