Learn More
Biolegend PE anti-human Integrin α9β1
Supplier:  Biolegend 351606
Encompass_Preferred
PE anti-human Integrin α9β1 [Y9A2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-671